1-Isocyanoadamantane,97% GC 1 * 0,1 g
Catalog # AMBDA708505-100MG
Supplier: AMBEED
Frequently Bought Together
Cyclohexane anhydrous, 99.5 % 1 * 1 L
Supplier: Merck
Isoamyl alcohol, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.001% H₂O) ≥99.8%
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.7%, CHROMASOLV™ Plus for HPLC, Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
N,N-Dimethylacetamide, anhydrous ≥99.8%
Supplier: Thermo Fisher Scientific
Acetone ≥99.0% Ph. Eur., Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Hydrochloric acid ≥36.5% ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
Ethanol (untaxed), anhydrous, tested according to Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Ethylene glycol monomethyl ether (2-methoxyethanol) 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran, anhydrous stabilised for grignard-reaction
Supplier: Thermo Fisher Scientific
Pyridine 99+%, extra pure
Supplier: Thermo Fisher Scientific
Hexane (mixture of isomers) 99% ACS
Supplier: Thermo Fisher Scientific