7-(2-Bromoethyl)-2-(furan-2-yl)-7H-pyrazolo[4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine,97% 1 * 0,1 g
Catalog # AMBDA918329-100MG
Supplier: AMBEED
7-(2-Bromoethyl)-2-(furan-2-yl)-7H-pyrazolo[4,3-e][1,2,4]triazolo[1,5-c]pyrimidin-5-amine,97% 1 * 0,1 g
Catalog # AMBDA918329-100MG
Supplier: AMBEED
Frequently Bought Together
n-Hexane, EMPLURA®, Supelco®
Supplier: Merck
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
2-Propanol ≥70% (v/v), SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
1,4-Dioxane ≥99% for HPLC
Supplier: Thermo Fisher Scientific
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Toluene ≥99.7%, CHROMASOLV™ for analysis of dioxins, furans and PCB, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Butanone (Methylethylketone) pure 1 * 2,5 L
Supplier: PanReac AppliChem
Ethanol eurodenatured ≥99%, GPR RECTAPUR® (1:1:1)
Supplier: VWR Chemicals
2-Propanol (USP, BP, Ph. Eur.) pure, pharma grade 1 * 200 L
Supplier: PanReac AppliChem
Hexane (mixture of isomers) ≥95%, Laboratory Reagent
Supplier: Honeywell Chemicals