N-Methyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetamide,95% 1 * 0,1 g
Catalog # AMBDA921311-100MG
Supplier: AMBEED
N-Methyl-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetamide,95% 1 * 0,1 g
Catalog # AMBDA921311-100MG
Supplier: AMBEED
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
m-Xylene for synthesis, Sigma-Aldrich®
Supplier: Merck
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrachloroethylene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Chlorobenzene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Propanol, EMPLURA®, Supelco®
Supplier: Merck
Dimethyl sulphoxide ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
p-Xylene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethanol absolute ≥99%
Supplier: KLINIPATH
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Acetone ≥99% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Supplier: Honeywell Chemicals