Methyl 5-bromo-2-((tert-butoxycarbonyl)amino)thiophene-3-carboxylate,95% 1 * 0,1 g
Catalog # AMBDA242443-100MG
Supplier: AMBEED
Methyl 5-bromo-2-((tert-butoxycarbonyl)amino)thiophene-3-carboxylate,95% 1 * 0,1 g
Catalog # AMBDA242443-100MG
Supplier: AMBEED
Frequently Bought Together
N,N-Dimethylacetamide anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
1,2,3,4-Tetrahydronaphthalene for synthesis, Sigma-Aldrich®
Supplier: Merck
Dimethyl sulfoxide EMPLURA 1 * 185 L
Supplier: Merck
Benzonitrile 99+% for spectroscopy
Supplier: Thermo Fisher Scientific
Chloroform ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Diisopropyl ether ≥98.5% (by GC) stabilised, Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol ≥99.5% NF, Ph. Eur., BP
Supplier: VWR Chemicals
Ethyl acetate ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Turpentine oil
Supplier: Roth Carl
Dimethyl sulphoxide, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Piperidine ≥99.5% (GC, on anhydrous basis) for peptide synthesis
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
ETHYL ACETATE 99.5% FOR SYNTHESIS 1 * 200 L
Supplier: PanReac AppliChem
Ethanolamine ACS
Supplier: Thermo Fisher Scientific