Sodium fluoride EMPROVE API Ph Eur,BP,USP 1 * 1 kg
Catalogus # 1.06441.0001
Supplier: MERCK PRODUCTION CHEMICALS
Sodium fluoride EMPROVE API Ph Eur,BP,USP 1 * 1 kg
Catalogus # 1.06441.0001
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Sodium tungstate dihydrate ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium nitrite ≥99.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Zinc iodide, EMSURE® for analysis, Supelco®
Supplier: Merck
Potassium hexacyanoferrate(II) trihydrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Ammonium nitrate ≥98.0%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium butyrate for synthesis, Sigma-Aldrich®
Supplier: Merck
Lanthanum(III) oxide 91%
Supplier: Thermo Fisher Scientific
Sodium sulphate, anhydrous 99+%, powder for analysis
Supplier: Thermo Fisher Scientific
Silver fluoride 98%
Supplier: Apollo Scientific
1-Heptanesulphonic acid sodium salt monohydrate ≥99%
Supplier: Thermo Fisher Scientific
Zirconium tetrachloride 99,5%
Supplier: Apollo Scientific
Potassium dihydrogen phosphate ≥99.0% (by acidimetry) ACS analytical reagent
Supplier: PanReac AppliChem
Sodium acetate trihydrate 99.5% for analysis
Supplier: Thermo Fisher Scientific