SPEX VAPLOCK PLUG, PFA, LUER PLUG X 1/4-28 UNF(M) FLAT BOTTOM 1 * 1 ST
Catalog # COLEGZ-12031-20
Supplier: Cole-Parmer
SPEX VAPLOCK PLUG, PFA, LUER PLUG X 1/4-28 UNF(M) FLAT BOTTOM 1 * 1 ST
Catalog # COLEGZ-12031-20
Supplier: Cole-Parmer
Frequently Bought Together
20Mm Borate Buffer Ph9.3 100Ml 1 * 100 mL
Supplier: Agilent
SPE columns, CHROMABOND® HLB
Supplier: MACHEREY-NAGEL
Kel-F® hub needles
Supplier: HAMILTON BONADUZ
Gastight® 1700 series syringes
Supplier: HAMILTON BONADUZ
Safety waste caps and ventilation kit, SymLine®
Supplier: S.C.A.T.
8MM PTFE SEPTUM, 1 * 100 items
Supplier: WATERS
Waste caps, b.safe
Supplier: Bohlender
Screw neck vials
Supplier: WATERS
Consumables for Thermo Scientific autosamplers
Supplier: Trajan Scientific and Medical
Reprosil Chiral-AM, 5 µm, 250 x 4.6 mm 1 * 1 items
Supplier: DR MAISCH
METAGUARD 2.0MM PURSUIT XRS 3U C18,3 1 * 1 items
Supplier: VARIAN
CORTECS C18 Column, 90Å, 2.7 µm, 4.6 mm X 100 mm 1 * 1 items
Supplier: WATERS
Avantor® ACE® C18, HPLC Columns, 3 µm
Supplier: Avantor