[EN]DI DIVOSAN ETHA-PLUS 6X0.75L 1 * 6 Fl
Catalog # DIVY101104303
Leverancier: Diversey
Frequently Bought Together
Ethyl acetate, Prepsolv® for preparative chromatography, Supelco®
Leverancier: Merck
N,N-Dimethylformamide, dehydrated (max. 0.015% H₂O) ≥99.9% for synthesis
Leverancier: VWR Chemicals
Hydrogen peroxide 35%, J.T.Baker®
Leverancier: Avantor
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
N,N-Dimethyl-p-toluidine for synthesis, Sigma-Aldrich®
Leverancier: Merck
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
2-Propanol ≥99.5% (by GC), EMPARTA® ACS analytical reagent, Supelco®
Leverancier: Merck
Toluene, anhydrous ≥99.8%
Leverancier: Thermo Fisher Scientific
n-Hexane, extra dry 96+%, AcroSeal®
Leverancier: Thermo Fisher Scientific
Water, Ultrapure for HPLC
Leverancier: Thermo Fisher Scientific
3-(Chloromethyl)benzoyl chloride 98%
Leverancier: Thermo Fisher Scientific
Cyclohexane ≥99%
Leverancier: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
Leverancier: VWR Chemicals
Toluene ≥99.8% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Leverancier: MACRON AVANTOR BRAND
Glycerine ≥99.0% (by GC), Reagent Grade
Leverancier: Honeywell Chemicals