Mercaptan Sulfur Set for UOP 163 and ASTM D3227: S @ 30 µg/g in 80% Isooctane/20% Toluene 1 * 1 SET
Catalog # PROOVHGUOP163-30-6
Supplier: LGC Standards PROMOCHEM
Mercaptan Sulfur Set for UOP 163 and ASTM D3227: S @ 30 µg/g in 80% Isooctane/20% Toluene 1 * 1 SET
Catalog # PROOVHGUOP163-30-6
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
CEFAZOLIN SODIUM SALT 1 * 75 mg
Supplier: Merck
Gold standard solution, for ICP, in 20% HCl, VHG
Supplier: LGC Standards PROMOCHEM
4-AMINOBENZYL ALCOHOL 1 * 100 mg
Supplier: Merck
Calcium Standard for AAS, TraceCERT TM 1 * 250 mL
Supplier: Merck
Barium standard for Ion Chromatography, (CRM), Fluka™
Supplier: Honeywell Chemicals
n-Pentadecane, Supelco®
Supplier: Merck
Pirimicarb Analytical standard, reference material, Fluka™
Supplier: Honeywell Chemicals
L-Citrulline, Reference Standard
Supplier: USP
PHOSPHONIC ACID 100µG/ML IN ACETONITRILE 1 * 1 mL
Supplier: EHRENSTORFER
Supelco® TraceCERT® Organic Reference Standard, Tebufenpyrad
Supplier: Merck
Toxaphene Parlar-No. 44 1 ng/µl 1 * 1 mL
Supplier: EHRENSTORFER
TOLUENE D8 1 * 0,1 g
Supplier: EHRENSTORFER
Buffer Solutions
Supplier: Thermo Fisher Scientific