[EN]RAPIDQC PLUS LEVEL 1,30 X 2.5 ML AMP 1 * 1 KIT
Catalog # BAYR301120
Supplier: BAYER VITAL
Frequently Bought Together
FIXATIVE CAL-RITE 1GAL 1 * 1 items
Supplier: Epredia
Rhodamine B base 97% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Reagents for molybdate determination using Lovibond® MD Photometers
Supplier: Lovibond
REAGENT KIT FOR PRIMELAB ID98 (50 TESTS) 1 * 1 items
Supplier: WATER ID PRIMELAB
Visual comparator test kits, silica
Supplier: Hach
trans-4-Hydroxycinnamic acid ≥98% (by HPLC), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
SALM O ANTIS FCT 19 1 * 1 Vial
Supplier: DIFCO BD
Nitrite VHR L, 500 ml
Supplier: Lovibond