Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Recycling solution for plastic packaging, Zero Waste Box™

Recycling solution for plastic packaging, Zero Waste Box™

Disposal box for plastic packaging, medium, 78 L, ≤15 kg, 305×305×840 mm
Catalog # 129-2556
   Product Transparency and Disclosure|Waste Reducing
Supplier:   TERRACYCLE
Recycling solution for plastic packaging, Zero Waste Box™
Catalog # 129-2556
Supplier:  TERRACYCLE
   Product Transparency and Disclosure|Waste Reducing
Recycling solution for plastic packaging, Zero Waste Box™
Recycling solution for plastic packaging, Zero Waste Box™

Disposal box for plastic packaging, medium, 78 L, ≤15 kg, 305×305×840 mm

Price
153,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

A Zero WasteBox™ is a complete and convenient solution which includes the storage, shipping and recycling of a particular type of waste that isn’t currently recycled through local councils or traditional recycling facilities.

  • Reduce the amount of waste that you send to landfill or incinerate
  • Recycle the non-recyclable and reduce your carbon footprint
  • Improving your organisation's sustainability can boost your corporate image and improve employee engagement and satifaction

Use this Zero WasteBox™ to recycle any flexible plastic packaging, carrier bags and shipping material including bubble wrap, polystyrene, airbags, shrink wrap, cushioning pallet liners and covers. Any hard / rigid plastic packaging and plastic containers. Use this box to recycle VWR® PureStep EnviroTack™ mat layers, including top and bottom release liners.

How it works;
Order - Select a Zero WasteBox™ based on what and how much you would like to recycle.
Collect - Fill your box with the accepted waste and seal the box with adhesive tape
Ship - Contact ups.com/nl, 020-5040500 or use the pre-paid UPS shipping label affixed to the back of your box, send your filled box to TerraCycle® for recycling. UPS requires a minimum of 24 hours’ notice and does pick ups Monday to Friday between 9am and 5pm. Pick-ups can be arranged online or by telephone. All shipments are carbon-neutral, meaning the carbon emissions released during transport will be offset.
Reorder - Continue to recycle and reorder a new Zero WasteBox™ . Your collected waste will find a second life as recycled materials, reducing the need to extract virgin materials and depleting resources.

Cardboard boxes are made from 84% recycled material. Box container and liner are 100% recyclable.

 : TerraCycle® will not landfill or incinerate any of the waste received.

 : Do not send in food content and liquid waste. Plant-based or biodegradable plastics (e.g. PLA). Any non-plastic waste: paper, metals, cardboard, glass. Plastic cards. Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic) which presents a danger to the environment, or to people. Batteries, pressurised canisters, broken glass and medical waste.

Specifications
  • Capacity:
    78 L
  • Description:
    Disposal box for plastic packaging, medium
  • Seq:
    02
  • W×D×H:
    305×305×840 mm
  • Load capacity:
    ≤15 kg
  • Cat. No.:
    129-2556
  • Environmental certification:
    FSC (Certificate code FSC: BV-COC-150648), Bureau Veritas, Third party verified LCA
  • Environmentally Preferable:
    Product Transparency and Disclosure|Waste Reducing