Frequently Bought Together
Ammonia solution 25%, TECHNICAL
Supplier: VWR Chemicals
Ammonium formate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Hydrogen peroxide 6% (w/v) (20 volumes), GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, peracetic acid, Dosatest
Supplier: VWR Chemicals
Ammonium acetate ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Kjeldahl tablets, Kjeltabs
Supplier: THOMPSON & CAPPER
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
Test strips, peroxide, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Test kits, photometric, Zinc, LCK and LCS cuvette tests
Supplier: Hach
BOND EPITOPE RETRIEVAL SOLUTION 2 1 * 1 L
Supplier: LEICA MICROSYSTEMS
VWR® Powder Packs for Determination of Copper, Reagents for Photometers
Supplier: VWR Chemicals
L(+)-Ascorbic acid
Supplier: Hach