Order Entry
Netherlands
ContactUsLinkComponent
Obj. HI PLAN I 20x/0.30 Objective HI PLAN I 20x/0.30 Free working distance: 3.7 - 2.4 mm For use with cover glass of 0 - 2 mm thickness Useful and recommended for Integrated Leica Modulation Contrast (IMC) 1 * 1 items
Catalogus # LEIM11506264
Supplier:  LEICA MICROSYSTEMS
Obj. HI PLAN I 20x/0.30 Objective HI PLAN I 20x/0.30 Free working distance: 3.7 - 2.4 mm For use with cover glass of 0 - 2 mm thickness Useful and recommended for Integrated Leica Modulation Contrast (IMC) 1 * 1 items
Catalogus # LEIM11506264
Supplier:  LEICA MICROSYSTEMS
Frequently Bought Together