Sodium sulfate anhydrous EMPROVE EXPERT Ph Eur,BP,ChP,USP 1 * 1 kg
Catalog # 1.28225.1001
Supplier: MERCK PRODUCTION CHEMICALS
Sodium sulfate anhydrous EMPROVE EXPERT Ph Eur,BP,ChP,USP 1 * 1 kg
Catalog # 1.28225.1001
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Sodium hydrogen carbonate ≥98%, TECHNICAL
Supplier: VWR Chemicals
Sodium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Potassium iodide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Potassium chloride 99.5-101.0%, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Sodium carbonate, anhydrous 99.5-100.5% NF, Ph. Eur., BP
Supplier: VWR Chemicals
Ammonium metavanadate ≥99.0% analytical reagent
Supplier: VWR Chemicals
Sodium chloride regeneration salt, TECHNICAL for water softening
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Potassium chromate ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Sodium salicylate ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
di-Potassium hydrogen phosphate, anhydrous ≥98.0%, EMPROVE® ESSENTIAL Ph. Eur., BP, E340, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS