ALLO-L-ISOLEUCINE (13C6 97-99%13C 15N 97-99 (Bulk Gram) Packaged 1 * 10 mg
Catalog # CAMBCNLM-8670-PK
Supplier: CAMBRIDGE ISOTOPE LABS
ALLO-L-ISOLEUCINE (13C6 97-99%13C 15N 97-99 (Bulk Gram) Packaged 1 * 10 mg
Catalog # CAMBCNLM-8670-PK
Supplier: CAMBRIDGE ISOTOPE LABS
Frequently Bought Together
Serological pipettes, Stripette™
Supplier: Corning
Eppendorf® Protein LoBind Plates
Supplier: EPPENDORF
Water, GPR RECTAPUR®
Supplier: VWR Chemicals
N,N-Dimethylformamide 99.0-100.00%, PESTINORM® for headspace gas chromatography
Supplier: VWR Chemicals
Glycerine ≥97%, TECHNICAL
Supplier: VWR Chemicals
Serological pipettes
Supplier: Greiner Bio-One
Glycerine (vegetable based), anhydrous, EMPROVE® EXPERT ACS, Ph. Eur., BP, USP, JP for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
n-Hexane, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
MicroWell™ Plates, 96 well, Nunc™
Supplier: Thermo Fisher Scientific
Spare Closures for VWR® Media Bottles
Supplier: VWR Collection
Isoamyl alcohol 98%, pure
Supplier: Thermo Fisher Scientific
Ethanol ≥96% (v/v), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
TRAY PP 540x435x130 MM 1 * 1 items
Supplier: SEMADENI
Petroleum spirit, 35…60 °C ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals