SIALIDASE TESTING PANEL (2-AB LABELLED) 1 * 100 Pmol
Catalog # LUDGCABSTPNEUAC-01
Supplier: LUDGER
Frequently Bought Together
NANOCOLOR® Photometric Tube Tests, Surfactants
Supplier: MACHEREY-NAGEL
EOSIN RAL 555 1L 1 * 1 L
Supplier: RAL
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Supplier: VWR Chemicals
Colour comparison disk Ammonium suitable for Cat. No. 920006 VISOCOLOR HE. 1 * 1 items
Supplier: MACHEREY-NAGEL
Anti-IGHG4 Human Antibody [clone: MOPC-21]
Supplier: Abcam
EOSIN RAL 555 2.5L 1 * 2,5 L
Supplier: RAL
Polyvinyl alcohol, fully hydrolysed, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
POTASSIUM IODURE 20% 1 * 1 L
Supplier: PROLABO CMP
PARAPLAST REGULAR BULK PACK 56°C 1 * 5 kg
Supplier: LEICA HISTOLOGY
Test paper, qualitative, Nitratesmo
Supplier: MACHEREY-NAGEL