2AB LABELLED CORE 1 O- GLYCAN, C1 1 * 100 Pmol
Catalog # LUDGCAB-C1-01
Supplier: LUDGER
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Water Reag. Ph. Eur. 1095506 nitrate-free
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach