FA2B glycan (G0B), 2-AA labelled 1 * 100 Pmol
Catalog # LUDGCAA-FA2B-01
Leverancier: LUDGER
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Leverancier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Leverancier: VWR Chemicals
VWR®, Sulphate 1000 mg/L in water standard for ion chromatography
Leverancier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Leverancier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Leverancier: Merck
Potassium pyroantimonate 0.05 mol/l in potassium hydroxide solution Reag. Ph. Eur. 1071301
Leverancier: VWR Chemicals
Nitrogen (total) CRM, Standard Solutions, Supelco®
Leverancier: Merck
VWR® Buffer Solutions, Secondary Standards
Leverancier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Leverancier: Merck
Conductivity standards, potassium chloride, referenced at 25 °C
Leverancier: Hach
Redox buffer solutions
Leverancier: Mettler - Toledo