Alpha-Gal standard (2-AA labelled) 1 * 100 Pmol
Catalog # LUDGCAAALPHAGAL-01
Supplier: LUDGER
Frequently Bought Together
Water Reag. Ph. Eur. 1095506 nitrate-free
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Starch 1% in aqueous solution stabilised Reag. Ph. Eur. 1085103
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
Supplier: VWR Chemicals
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
VWR®, pH electrode maintenance kit
Supplier: VWR Chemicals
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond