A2G1 glycan (G1), 2AA labelled 1 * 100 Pmol
Catalog # LUDGCAA-A2G1-01
Supplier: LUDGER
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
VWR® ARISTAR®, Antimony Standard Solutions for ICP-MS
Supplier: VWR Chemicals
Pesticide reference standard, Aldicarb 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Supplier: EHRENSTORFER
Chromium standard solution, 1000 mg/L Cr in 5% hydrochloric acid (from Cr), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Total organic carbon (TOC) standards
Supplier: REAGECON
Dalfampridine Related Compounds, Reference Standard
Supplier: USP
VWR®, Distillation standards for the petroleum industry, Certified Reference Materials
Supplier: VWR Chemicals
Pymetrozine certified 1 * 0,1 g
Supplier: EHRENSTORFER
Sulfamethoxazole, Reference Standard
Supplier: USP
FREEZING POINT STD CRYOSCOPE 621 100ML 1 * 1 items
Supplier: REAGECON
Supelco® Organic Reference Standard, (+)-Menthofuran
Supplier: Merck
Mix of 6 quantitative monosaccharide standards 1 * 60 Nmo
Supplier: LUDGER
Eicosapentaenoic Acid, Reference Standards
Supplier: USP
Cadusafos
Supplier: EHRENSTORFER
Ethyl 4-hydroxybenzoate
Supplier: EDQM