IgG N-glycan library (procainamide) 1 * 50 Pmol
Catalog # LUDGCPROC-IGG-02
Supplier: LUDGER
Frequently Bought Together
pH buffer solutions
Supplier: Mettler - Toledo
Conductivity standards, Certipur® Supelco®
Supplier: Merck
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Chloride 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ