NA3 glycan (A3G3), procainamide labelled 1 * 20 Pmol
Catalog # LUDGCPROC-NA3-01
Supplier: LUDGER
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Buffer solutions
Supplier: Hanna
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Chloride Standard for IC 1 * 100 mL
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
Reagents for chloride determination using Lovibond® MD photometers
Supplier: Lovibond
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
ICP METAL STANDARD 31 COMPONENT 1 * 100 mL
Supplier: CPACHEM
pH Standard Solutions, IUPAC, Certified
Supplier: Hach