Man-7 glycan, procainamide labelled 1 * 20 Pmol
Catalog # LUDGCPROC-MAN7-01
Supplier: LUDGER
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® ARISTAR®, Multi-Element Calibration Standard IV in dil. Nitric Acid for ICP
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Calcium carbonate, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Buffer solutions as per DIN 19266-01: 2000
Supplier: WTW
Benzoic acid, Supelco®
Supplier: Merck
pH Standard Solutions, IUPAC, Certified
Supplier: Hach