Man-5 glycan, procainamide labelled 1 * 20 Pmol
Catalog # LUDGCPROC-MAN5-01
Supplier: LUDGER
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Sulfate standard solution
Supplier: Hach
Supelco® PESTANAL® Organic Reference Standard, Daminozide
Supplier: Merck
Supelco® PESTANAL® Organic Reference Standard, Tebuconazole
Supplier: Merck
Supelco® TraceCERT® Organic Reference Standard, Imidacloprid
Supplier: Merck
SILDENAFIL 1 * 10 mg
Supplier: LGC Standards PROMOCHEM
Organic reference standard, 4-Octylphenol 10 µg/ml in isooctane, Dr Ehrenstorfer
Supplier: EHRENSTORFER
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
STANDARD AU 4MG/L IN HNO3 5% HCL 15% 1 * 500 mL
Supplier: CUSTOM MADE CHEMICALS LAB
Tridemorph
Supplier: EHRENSTORFER
Isoxaflutole certified 1 * 0,1 g
Supplier: EHRENSTORFER
pH buffer solutions, colour coded
Supplier: Hach
Tefuryltrione 1 * 50 mg
Supplier: EHRENSTORFER