IGG N-GLYCAN LIBRARY (PERMETHYLATED) 1 * 20 Tests
Catalog # LUDGCPM-IGG-01
Supplier: LUDGER
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Nitrogen (total) CRM, Standard Solutions, Supelco®
Supplier: Merck
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Potassium pyroantimonate 0.05 mol/l in potassium hydroxide solution Reag. Ph. Eur. 1071301
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond