MANNOSE 8 QUANTITATIVE STANDARD 1 * 1 EA
Catalog # LUDGBQ-CN-MAN8-10U
Supplier: LUDGER
Frequently Bought Together
Geleidbaarheidsstandaarden, Certipur®, Supelco®
Supplier: Merck
Bufferoplossingen, Certipur® met kleurcode, Supelco®
Supplier: Merck
VWR® ARISTAR®, Multi-Element Calibration Standard IV in dil. Nitric Acid for ICP
Supplier: VWR Chemicals
VWR® AVS TITRINORM, Bufferoplossingen, 20 °C
Supplier: VWR Chemicals
Standaarden
Supplier: ROTRONIC INSTRUMENTS
Buffer solutions
Supplier: Mettler - Toledo
Standaardoplossingen voor geleidbaarheid
Supplier: VWR Chemicals
pH-buffers, klaar voor gebruik
Supplier: WTW
pH Standard Solutions, IUPAC, Certified
Supplier: Hach