Chitotriose quantitative standard (unlabelled) 1 * 5 Nmo
LUDGBQCHITOTRIOSE1
: LUDGER
Frequently Bought Together
pH buffers, DuraCal
: HAMILTON BONADUZ
di-Ammonium oxalate solution Reag. Ph. Eur. 1005901
: VWR Chemicals
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffer solutions
: Mettler - Toledo
Buffer solutions in sachets
: Mettler - Toledo
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
: HAMILTON BONADUZ