Ludger BioQuant GPEP A2G2S2 glycopeptide standard 1 * 3 Nmo
Catalog # LUDGBQGPEPA2G2S210
Supplier: LUDGER
Ludger BioQuant GPEP A2G2S2 glycopeptide standard 1 * 3 Nmo
Catalog # LUDGBQGPEPA2G2S210
Supplier: LUDGER
Frequently Bought Together
Conductivity standards, Certipur® Supelco®
Supplier: Merck
VWR®, pH electrode maintenance kit
Supplier: VWR Chemicals
BUFFER PH 4,01 DOSING BOTTLE 1 * 500 mL
Supplier: HAMILTON BONADUZ
pH buffers, ready to use
Supplier: WTW
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Accessories for chloride analyser, model 926
Supplier: SHERWOOD SCIENTIFIC INSTRUMENT
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, according to EP and USP, Certipur®
Supplier: Merck
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion
pH Standard Solutions, IUPAC, Certified
Supplier: Hach