Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT PNGASE F 1 * 60 Tests
Catalog # LUDGE-RPNG01
Supplier:
LUDGER
Expand image
RECOMBINANT PNGASE F 1 * 60 Tests
Catalog # LUDGE-RPNG01
Supplier:
LUDGER
Frequently Bought Together
DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
Ligation Sequencing Kits
Supplier: Oxford Nanopore Technologies
FlashGel™ DNA system
Supplier: LONZA
PathHunter eXpress mGLP1R CHO-K1 β-Arrestin GPCR Assay 1 * 1 KIT
Supplier: DISCOVERX
Genomic DNA ScreenTape Analysis (ScreenTape and Reagents)
Supplier: AGILENT
Rapid sequencing kits
Supplier: Oxford Nanopore Technologies
cAMP Hunter eXpress PTGDR CHO-K1 GPCR Assay 1 * 1 KIT
Supplier: DISCOVERX
Qualitative DNA Kits
Supplier: AGILENT