Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Sample Vials, with Snap-Cap

Sample Vials, with Snap-Cap

30,00 ml, 28×75 mm, Glass, Amber, Yes, Snap cap, closed top, 28 mm, PE, White, -
Catalog # HECH42786020
Sample Vials, with Snap-Cap
Catalog # HECH42786020
Sample Vials, with Snap-Cap
Sample Vials, with Snap-Cap

30,00 ml, 28×75 mm, Glass, Amber, Yes, Snap cap, closed top, 28 mm, PE, White, -

Price
107,00 €
Log in for availability
1 * 200 ST
Product Details & Documents

Soda-lime glass, amber.

  • Amber glass especially for light sensitive liquids
  • White snap-caps are impervious to ultraviolet light
  • Snap-caps included

Ideal for storing medication as liquid, powder or capsules.

Specifications
  • Cap included:
    Yes
  • Cap material:
    PE
  • Cap Size:
    28 mm
  • Ø×H:
    28×75 mm
  • Sterile:
    -
  • Capacity:
    30,00 ml
  • Colour:
    Amber
  • Material:
    Glass
  • Seq:
    04
  • Cap type:
    Snap cap, closed top
  • Cap colour:
    White
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.