n-Butyl methacrylate, 99%, stab. with 15-20 ppm 4-methoxyphenol 1 * 180 kg
Catalog # A19068.PG
Supplier: Thermo Fisher Scientific
n-Butyl methacrylate, 99%, stab. with 15-20 ppm 4-methoxyphenol 1 * 180 kg
Catalog # A19068.PG
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Pyrogallol for synthesis, Sigma-Aldrich®
Supplier: Merck
Methanol with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Butanone (Methylethylketone) pure 1 * 2,5 L
Supplier: PanReac AppliChem
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
2,2,4-Trimethylpentane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Supplier: Honeywell Chemicals
Dimethyl Sulfoxide (USP, BP, Ph. Eur.) pharma grade 1 * 190 L
Supplier: PanReac AppliChem
Bromoform 99+% stabilised
Supplier: Thermo Fisher Scientific
Dichloromethane 99.9% stabilised, AcroSeal® for biochemistry
Supplier: Thermo Fisher Scientific
Formamide 99.5+%, extra pure
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
2-Propanol, BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor