7-FLUORO-3-(METHOXYMETHOXY)-8-(2-TRIISOPROPYLSILYLETHYNYL)NAPHTHALEN-1-OL 1 * 100 g
PHBLPBU9519-100G
7-FLUORO-3-(METHOXYMETHOXY)-8-(2-TRIISOPROPYLSILYLETHYNYL)NAPHTHALEN-1-OL 1 * 100 g
PHBLPBU9519-100G
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Ethanol eurodenatured ≥96%, TechniSolv® (1:1:1)
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals