KAGER TMC IRREVERSIBLE TEMPERATURANZEIGE 1 * 250 mL
Catalog # KAGEMC153-14
Supplier: KAGER
Frequently Bought Together
UltraClear® for histology, J.T.Baker®
Supplier: Avantor
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Potassium hydroxide 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips for peracetic acid, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 50% in aqueous solution, TECHNICAL
Supplier: VWR Chemicals
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test kits, ammonium, Spectroquant®, Supelco®
Supplier: Merck
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Ammonia solution ∼32%, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
Acid Red 183 30% (dye content), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach