Polink-2 Plus HRP Polymer and AP polymer detection for HUMAN antibody on ANIMAL tissue 1 * 6 mL
Catalog # ORIGD88-6
Supplier: OriGene
Polink-2 Plus HRP Polymer and AP polymer detection for HUMAN antibody on ANIMAL tissue 1 * 6 mL
Catalog # ORIGD88-6
Supplier: OriGene
Frequently Bought Together
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Transfer pipettes
Supplier: RATIOLAB
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection