CUSTOM SOLUTION SACCCHARIN 10µG/ML METH 1 * 10 mL
Catalog # PROOSL25560
Supplier: LGC Standards PROMOCHEM
CUSTOM SOLUTION SACCCHARIN 10µG/ML METH 1 * 10 mL
Catalog # PROOSL25560
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
Conductivity standards, KCl, referenced at 25 °C
Supplier: REAGECON
HALOPERIDOL FOR PEAK IDENTIFICATION 1 * 25 mg
Supplier: EDQM
Supelco® VETRANAL® Organic Reference Standard, Doramectin
Supplier: Merck
Thiazafluron certified 1 * 0,25 g
Supplier: EHRENSTORFER
1,4-SORBITAN 1 * 500 mg
Supplier: Merck
HIGH MANNOSE GLYCAN STANDARD LIBRARY 1 * 10 µG
Supplier: LUDGER
CHEM013 MEASUREMENT STANDARD
Supplier: MARKET SOURCE PART PROCESS
Supelco® Organic Reference Standard, Apigenin
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Supplier: VWR Chemicals
Dinoseb acetate
Supplier: EHRENSTORFER
Total Cyanide-WP, (CRM), Supelco®
Supplier: Merck
Standards for water pollution QC check, inorganics, VHG
Supplier: LGC Standards PROMOCHEM
Ciprofloxacin Hydrochloride, Reference Standard
Supplier: USP
Trace Metals 1 in Seawater - Whole Volume, (CRM), Supelco®
Supplier: Merck
TRICLABENDAZOLE FOR SYSTEM SUITABILITY 1 * 0,12 mg
Supplier: EDQM
Bis(2-ethylhexyl)phthalate
Supplier: EHRENSTORFER