Recombinant protein of human cancer/testis antigen 1B (CTAG1B/NY-ESO-1) 1 * 20 µG
Catalog # ORIGTP313318
Supplier: OriGene
Recombinant protein of human cancer/testis antigen 1B (CTAG1B/NY-ESO-1) 1 * 20 µG
Catalog # ORIGTP313318
Supplier: OriGene
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR® Blender Bags
Supplier: VWR Collection
Decontamination solution, DNA AWAY®
Supplier: Thermo Fisher Scientific
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
Cell culture flasks, Falcon®
Supplier: Corning
Peqlab peqGOLD, Agarose, Universal
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
E.Z.N.A.® Plasmid Mini kit I
Supplier: OMEGA BIO-TEK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
Bioanalyzer RNA Analysis
Supplier: AGILENT
RNA ScreenTape Analysis
Supplier: AGILENT