TRACE ELEMENTS FORTIFIED SAMPLE 1 * 250 mL
Catalog # PROONWTMDA-54.6
Supplier: LGC Standards PROMOCHEM
TRACE ELEMENTS FORTIFIED SAMPLE 1 * 250 mL
Catalog # PROONWTMDA-54.6
Supplier: LGC Standards PROMOCHEM
Frequently Bought Together
PRIMAQUINE PHOSPHATE 1 * 500 mg
Supplier: Merck
α-Ketoglutaric acid disodium salt hydrate, Supelco®
Supplier: Merck
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
Ferric chloride-sulphamic acid reagent Reag. Ph. Eur. 1037804
Supplier: VWR Chemicals
Buffer Solution pH 9.23 (20°C) 1 * 1.000 mL
Supplier: PanReac AppliChem
Buffer solutions in sachets, perpHect® Pouches, Orion™
Supplier: Thermo Orion
PRALLETHRIN 1 * 100 mg
Supplier: EHRENSTORFER
2,4,4-Trimethyl-1-pentene
Supplier: EHRENSTORFER
SPE sorbents, CHROMABOND®
Supplier: MACHEREY-NAGEL
2-Butanol, Reference Standard
Supplier: USP
Supelco® PESTANAL® Organic Reference Standard, λ-Cyhalothrin
Supplier: Merck
DL-α-Tocopherol acetate
Supplier: EDQM