Buffer solution, pH 12.45 (+0.01 25oC), No Color, Specpurer, NIST Traceable 1 * 4 L
Catalog # 47322.K7
Supplier: Thermo Fisher Scientific
Buffer solution, pH 12.45 (+0.01 25oC), No Color, Specpurer, NIST Traceable 1 * 4 L
Catalog # 47322.K7
Supplier: Thermo Fisher Scientific
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Diethyl ether ≥99.7% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
Supplier: VWR Chemicals