Hirudin (54-65 non-sulfated) (trifluoroacetate salt) 1 * 25 mg
Catalog # CAYM33716-25
Supplier: Cayman Chemical
Hirudin (54-65 non-sulfated) (trifluoroacetate salt) 1 * 25 mg
Catalog # CAYM33716-25
Supplier: Cayman Chemical
Frequently Bought Together
Dimethyl sulphoxide, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
n-Hexane, EMPLURA®, Supelco®
Supplier: Merck
N-HEPTANE FOR LIQUID CHROMATOGRAPHY 1 * 185 L
Supplier: Merck
Ethanolamine ACS
Supplier: Thermo Fisher Scientific
Glycerine, Hi-LR™
Supplier: HIMEDIA
1-Chlorobutane ≥99.5% for HPLC
Supplier: Thermo Fisher Scientific
Water with 0,1% (v/v) acetic acid, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Chloroform ≥99.8% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Bromoform 99+% stabilised
Supplier: Thermo Fisher Scientific
n-Hexane 95%, ChromAR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Petroleum Ether 40-60C for pesticide analysis 1 * 2,5 L
Supplier: PanReac AppliChem
Toluene (Reag. Ph. Eur.) for analysis, ACS, ISO 1 * 2,5 L
Supplier: PanReac AppliChem
N,N-DIMETHYLFORMAMIDE AR | 2,5 DM3 1 * 2,5 L
Supplier: MACRON AVANTOR BRAND