100 mL PharmaCount™ 2M, 70µm Number of particles: 2000 ± 200 m
Catalog # MSPP2M070-100
Supplier: MARKET SOURCE PART PROCESS
100 mL PharmaCount™ 2M, 70µm Number of particles: 2000 ± 200 m
Catalog # MSPP2M070-100
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
Toluene ≥98%, TECHNICAL
Supplier: VWR Chemicals
Solvent ASTM D2896, Chlorobenzene/Acetic acid-mixture (2/1 v/v), TBN solvent, VOLUSOL®
Supplier: VWR Chemicals
Ethanol absolute 99.8% for HPLC
Supplier: Thermo Fisher Scientific
Iodomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Petroleum spirit, 80…110 °C
Supplier: Roth Carl
n-Pentane, anhydrous 99+%, extra pure
Supplier: Thermo Fisher Scientific
(±)-1,2-Propanediol ACS
Supplier: Thermo Fisher Scientific
Xylene (mixture of isomers), BAKER, J.T.Baker®
Supplier: Avantor
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
1,2-Dichloroethane 99+%, pure
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), liquid USP, BP, Ph. Eur., pure
Supplier: PanReac AppliChem
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Supplier: Merck
N,N-Dimethylacetamide 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Cyclohexane ≥99.5% (by GC), Puriss. p.a. ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Ethylbenzene 99.8%, pure
Supplier: Thermo Fisher Scientific