Order Entry
Netherlands
ContactUsLinkComponent
Precision laboratory thermometer enclosed scale, Capillaries form: transparent prismatic, liquid filling: spec.red, range: - 10+200C in 1C, total length: 350 mm, immersion length: total, including works certification with 3 calibration points. 1 * 1 items
Catalog # SCNL1042350/03/3P
Precision laboratory thermometer enclosed scale, Capillaries form: transparent prismatic, liquid filling: spec.red, range: - 10+200C in 1C, total length: 350 mm, immersion length: total, including works certification with 3 calibration points. 1 * 1 items
Catalog # SCNL1042350/03/3P
Frequently Bought Together