Order Entry
Netherlands
ContactUsLinkComponent
External measuring instrument with additional measuring head for temperature and humidity measurement. Product information on demand 1 * 1 items
Catalog # MEMMB04714IPP260EC
Supplier:  MEMMERT
External measuring instrument with additional measuring head for temperature and humidity measurement. Product information on demand 1 * 1 items
Catalog # MEMMB04714IPP260EC
Supplier:  MEMMERT
Frequently Bought Together