Order Entry
Netherlands
ContactUsLinkComponent
Human CD3E&CD3G Heterodimer Protein, His Tag&Tag Free (CDG-H52W5) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # P07766-1 (CD3E) & P09693-1 (CD3G)). 1 * 50 µG
Catalog # ACRBCDG-H52W5-50UG
Supplier:  ACRO Biosystems
Human CD3E&CD3G Heterodimer Protein, His Tag&Tag Free (CDG-H52W5) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # P07766-1 (CD3E) & P09693-1 (CD3G)). 1 * 50 µG
Catalog # ACRBCDG-H52W5-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together