Order Entry
Netherlands
ContactUsLinkComponent
SARS-CoV-2 S protein (K417N,D614G), His Tag (SPN-C52Hm) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). The mutations (K417N, D614G) were identified in the SARS-CoV-2 variant (known as B.1.351 o 1 * 500 µG
Catalog # ACRBSPN-C52HM500UG
Supplier:  ACRO Biosystems
SARS-CoV-2 S protein (K417N,D614G), His Tag (SPN-C52Hm) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). The mutations (K417N, D614G) were identified in the SARS-CoV-2 variant (known as B.1.351 o 1 * 500 µG
Catalog # ACRBSPN-C52HM500UG
Supplier:  ACRO Biosystems
Frequently Bought Together