Order Entry
Netherlands
ContactUsLinkComponent
SARS-CoV-2 S1 protein, Fc Tag (S1D-C5256) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # Q HD43416.1). The mutations (L18F, D80A, D215G, LAL242-244del, R246I, K417N, E484K, N501Y, D614G) were identified in th 1 * 100 µG
Catalog # ACRBS1D-C5256100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 S1 protein, Fc Tag (S1D-C5256) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # Q HD43416.1). The mutations (L18F, D80A, D215G, LAL242-244del, R246I, K417N, E484K, N501Y, D614G) were identified in th 1 * 100 µG
Catalog # ACRBS1D-C5256100UG
Supplier:  ACRO Biosystems
Frequently Bought Together