Order Entry
Netherlands
ContactUsLinkComponent
SARS-CoV-2 Spike S1, His Tag (S1N-C52Ht) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QH D43416.1). The mutations (T95I, G142D, E154K, L452R, E484Q, D614G, P681R) were identified in the SARS-CoV-2 variants 1 * 100 µG
Catalog # ACRBS1N-C52HT100UG
Supplier:  ACRO Biosystems
SARS-CoV-2 Spike S1, His Tag (S1N-C52Ht) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QH D43416.1). The mutations (T95I, G142D, E154K, L452R, E484Q, D614G, P681R) were identified in the SARS-CoV-2 variants 1 * 100 µG
Catalog # ACRBS1N-C52HT100UG
Supplier:  ACRO Biosystems
Frequently Bought Together