Order Entry
Netherlands
ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (SPN-C82Ed) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). The mutations (P26S, HV69-70del, V126A, Y144del, LLA241-243del, H245Y, S477N, E484K, T 1 * 200 µG
Catalogus # ACRBSPN-C82ED200UG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag (SPN-C82Ed) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). The mutations (P26S, HV69-70del, V126A, Y144del, LLA241-243del, H245Y, S477N, E484K, T 1 * 200 µG
Catalogus # ACRBSPN-C82ED200UG
Supplier:  ACRO Biosystems
Frequently Bought Together