Order Entry
Netherlands
ContactUsLinkComponent
Mouse ITGA11&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-M52W5) is expressed from human 293 cells (HEK293). It contains AA Phe 23 - Pro 1141 (ITGA11) & Gln 21 - Asp 728 (ITGB1) (Accession # P61622-1 (ITGA11) & P09055-1 (ITGB1)). 1 * 100 µG
Catalog # ACRBIT1-M52W5100UG
Supplier:  ACRO Biosystems
Mouse ITGA11&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-M52W5) is expressed from human 293 cells (HEK293). It contains AA Phe 23 - Pro 1141 (ITGA11) & Gln 21 - Asp 728 (ITGB1) (Accession # P61622-1 (ITGA11) & P09055-1 (ITGB1)). 1 * 100 µG
Catalog # ACRBIT1-M52W5100UG
Supplier:  ACRO Biosystems
Frequently Bought Together