Order Entry
Netherlands
ContactUsLinkComponent
Biotinylated Human 4-1BB (87-186), His,Avitag (41B-H82E4) is expressed from human 293 cells (HEK293). It contains AA Asp 87 - Gln 18 6 (Accession # Q07011-1). 1 * 200 µG
Catalog # ACRB41B-H82E4200UG
Supplier:  ACRO Biosystems
Biotinylated Human 4-1BB (87-186), His,Avitag (41B-H82E4) is expressed from human 293 cells (HEK293). It contains AA Asp 87 - Gln 18 6 (Accession # Q07011-1). 1 * 200 µG
Catalog # ACRB41B-H82E4200UG
Supplier:  ACRO Biosystems
Frequently Bought Together