Order Entry
Netherlands
ContactUsLinkComponent
Biotinylated Mouse PSCA, His,Avitag (PSA-M82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Asn 95 (Accessio n # P57096-1). 1 * 200 µG
Catalog # ACRBPSA-M82E3200UG
Supplier:  ACROBIOSYSTEMS
Biotinylated Mouse PSCA, His,Avitag (PSA-M82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 21 - Asn 95 (Accessio n # P57096-1). 1 * 200 µG
Catalog # ACRBPSA-M82E3200UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together