Order Entry
Netherlands
ContactUsLinkComponent
Rat ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-R52W9) is expressed from human 293 cells (HEK293). It contains AA Tyr 27 - Gly 1130 (ITGA2) & Gln 21 - Asp 729 (ITGB1) (Accession # A0A0G2K470-1 (ITGA2) & P49134-1 (ITGB1)). 1 * 1 mg
  ACRBIT1-R52W9-1MG
Rat ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-R52W9) is expressed from human 293 cells (HEK293). It contains AA Tyr 27 - Gly 1130 (ITGA2) & Gln 21 - Asp 729 (ITGB1) (Accession # A0A0G2K470-1 (ITGA2) & P49134-1 (ITGB1)). 1 * 1 mg
  ACRBIT1-R52W9-1MG
Frequently Bought Together